5-Pyridin-2-yl-1,3,4-oxadiazol-2-amine 95%
Dodavatel: Apollo Scientific
Synonyms:
5-(Pyridin-2-yl)-1,3,4-oxadiazol-2-amine
OR922722-1G
APOSOR922722-1GEA
5430
CZK
APOSOR922722-1G
5-Pyridin-2-yl-1,3,4-oxadiazol-2-amine 95%
5-Pyridin-2-yl-1,3,4-oxadiazol-2-amine
Vzorec:
C₇H₆N₄O MW: 162,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01710636 CAS číslo: 5711-72-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...