Tisk…

1-Cyclobutylpiperidin-4-ol 95%

Dodavatel: Apollo Scientific

OR922650-1G OR922650-250MG
APOSOR922650-1GEA 24170 CZK
APOSOR922650-1G APOSOR922650-250MG
1-Cyclobutylpiperidin-4-ol 95%
1-Cyclobutylpiperidin-4-ol
Vzorec: C₉H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD11878581
CAS číslo: 869224-62-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR