Decahydro-2-naphthol (mixture of isomers) 95%
Dodavatel: Apollo Scientific
OR922613-100G
OR922613-25G
APOSOR922613-100GEA
7670
CZK
APOSOR922613-100G
APOSOR922613-25G
Decahydro-2-naphthol (mixture of isomers) 95%
Decahydro-2-naphthol (mixture of isomers)
Vzorec:
C₁₀H₁₈O Storage Temperature: Ambient |
MDL Number:
MFCD00004132 CAS číslo: 825-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...