Tisk…

3-Methyl-2-butenyl benzoate 95%

Dodavatel: Apollo Scientific

Synonyms: Benzoic Acid 3-Methyl-2-butenyl Ester, Prenyl Benzoate, 3-Methylbut-2-en-1-yl benzoate, Benzoic Acid Prenyl Ester

OR922563-25G OR922563-5G
APOSOR922563-25GEA 7610 CZK
APOSOR922563-25G APOSOR922563-5G
3-Methyl-2-butenyl benzoate 95%
3-Methyl-2-butenyl benzoate
Vzorec: C₁₂H₁₄O₂
MW: 190,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036570
CAS číslo: 5205-11-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR