6-Chloro-3-oxo-2,3-dihydropyridazine-4-carboxylic acid methyl ester 95%
Dodavatel: Apollo Scientific
Synonyms:
Methyl 6-chloro-3-oxo-2,3-dihydropyridazine-4-carboxylate
OR922503-1G
OR922503-250MG
APOSOR922503-1GEA
3970
CZK
APOSOR922503-1G
APOSOR922503-250MG
6-Chloro-3-oxo-2,3-dihydropyridazine-4-carboxylic acid methyl ester 95%
6-Chloro-3-oxo-2,3-dihydropyridazine-4-carboxylic acid methyl ester
Vzorec:
C₆H₅ClN₂O₃ MW: 188.57 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16250585 CAS číslo: 89581-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...