Tisk…

5-Chloro-3-methyl-1H-indole-2-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR922498-1G OR922498-5G
APOSOR922498-1GEA 3940 CZK
APOSOR922498-1G APOSOR922498-5G
5-Chloro-3-methyl-1H-indole-2-carbaldehyde 95%
5-Chloro-3-methyl-1H-indole-2-carbaldehyde
Vzorec: C₁₀H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD08059791
CAS číslo: 40731-16-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR