3-(2-Methylallyloxy)benzaldehyde 95%
Dodavatel: Apollo Scientific
OR922489-1G
OR922489-5G
APOSOR922489-1GEA
6220
CZK
APOSOR922489-1G
APOSOR922489-5G
3-(2-Methylallyloxy)benzaldehyde 95%
3-(2-Methylallyloxy)benzaldehyde
Vzorec:
C₁₁H₁₂O₂ MW: 176.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07392462 CAS číslo: 38002-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...