Tisk…

2-(2-Pyridyl)ethanesulphonic acid 95%

Dodavatel: Apollo Scientific

OR922420-25G OR922420-5G
APOSOR922420-25GEA 10980 CZK
APOSOR922420-25G APOSOR922420-5G
2-(2-Pyridyl)ethanesulphonic acid 95%
2-(2-Pyridyl)ethanesulphonic acid
Vzorec: C₇H₉NO₃S
MW: 187,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006365
CAS číslo: 68922-18-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR