Tisk…

(2,4-Xylyloxy)acetic acid 95%

Dodavatel: Apollo Scientific

Synonyms: (2,4-Dimethylphenoxy)acetic acid, 2-(2,4-Dimethylphenoxy)acetic acid

OR922387-5G
APOSOR922387-5GEA 2440 CZK
APOSOR922387-5G
(2,4-Xylyloxy)acetic acid 95%
(2,4-Xylyloxy)acetic acid
Vzorec: (C₆H₃(CH₃)₂)OCH₂COOH
MW: 180.2
Storage Temperature: Ambient
MDL Number: MFCD00014355
CAS číslo: 13334-49-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR