Octylphosphonic acid 97%
Dodavatel: Apollo Scientific
Synonyms:
OPA, 1-Octylphosphonic acid, n-Octylphosphonic acid
OR922324-100G
OR922324-25G
OR922324-1KG
APOSOR922324-100GEA
7410
CZK
APOSOR922324-100G
APOSOR922324-25G
APOSOR922324-1KG
Octylphosphonic acid 97%
Octylphosphonic acid
Vzorec:
C₈H₁₉O₃P Bod tání: 93…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015841 CAS číslo: 4724-48-5 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...