Tisk…

Isobutyryl bromide 95%

Dodavatel: Apollo Scientific

OR922323-25G OR922323-5G
APOSOR922323-25GEA 14290 CZK
APOSOR922323-25G APOSOR922323-5G
Isobutyryl bromide 95%
Isobutyryl bromide
Vzorec: C₄H₇BrO
Storage Temperature: Ambient
MDL Number: MFCD00059468
CAS číslo: 2736-37-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR