Tisk…

Phenyl-3-chloropropionate 95%

Dodavatel: Apollo Scientific

Synonyms: Phenyl 3-chloropropanoate, 3-Chloropropionic Acid Phenyl Ester

OR922263-100G
APOSOR922263-100GEA 3060 CZK
APOSOR922263-100G
Phenyl-3-chloropropionate 95%
Phenyl-3-chloropropionate
Vzorec: C₉H₉ClO₂
MW: 184,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045298
CAS číslo: 24552-27-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR