Zinc formate 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR922222-25G
OR922222-100G
OR922222-500G
APOSOR922222-25GEA
827
CZK
APOSOR922222-25G
APOSOR922222-100G
APOSOR922222-500G
Zinc formate 95+%
Zinc formate
Vzorec:
C₂H₂O₄Zn MW: 155,40 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00050836 CAS číslo: 557-41-5 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...