2-(4-Ethoxyphenyl)-2-methylpropanol 95%
Dodavatel: Apollo Scientific
OR922148-25G
OR922148-5G
APOSOR922148-25GEA
4570
CZK
APOSOR922148-25G
APOSOR922148-5G
2-(4-Ethoxyphenyl)-2-methylpropanol 95%
2-(4-Ethoxyphenyl)-2-methylpropanol
Vzorec:
C₁₂H₁₈O₂ MW: 194,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07787279 CAS číslo: 83493-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...