Tisk…

2-Methyl-2H-indazole-3-methanol 95%

Dodavatel: Apollo Scientific
Varování

OR922120-1G OR922120-250MG
APOSOR922120-1GEA 1620 CZK
APOSOR922120-1G APOSOR922120-250MG
2-Methyl-2H-indazole-3-methanol 95%
2-Methyl-2H-indazole-3-methanol
Vzorec: C₉H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD08060471
CAS číslo: 58536-48-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR