Bis(2-chlorethyl)ether 95%
Dodavatel: Apollo Scientific
Synonyms:
2-Chloroethyl ether
OR922075-100G
OR922075-1KG
OR922075-500G
APOSOR922075-100GEA
3110
CZK
APOSOR922075-100G
APOSOR922075-1KG
APOSOR922075-500G
Bis(2-chlorethyl)ether 95%
Bis(2-chlorethyl)ether
Vzorec:
C₄H₈Cl₂O MW: 143.01 Bod varu: 178 °C/760mmHg Bod tání: –52 °C Hustota: 1.22 g/cm³ (25 °C) Bod vzplanutí: 55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000975 CAS číslo: 111-44-4 UN: 1916 ADR: 6.1+3 II Merck Index: 13,03091 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...