Tisk…
Ethylene glycol monoisopropyl ether 95%
Dodavatel: Apollo Scientific
Synonyms: 2-Isopropoxyethanol
OR922073-100G OR922073-1KG OR922073-500G
APOSOR922073-100GEA 3110 CZK
APOSOR922073-100G APOSOR922073-1KG APOSOR922073-500G
Ethylene glycol monoisopropyl ether 95%
Ethylene glycol monoisopropyl ether
Vzorec: C₅H₁₂O₂
MW: 104.15
Storage Temperature: Ambient
MDL Number: MFCD00002866
CAS číslo: 109-59-1