Tisk…

2-Ethylphenyl acetate 95%

Dodavatel: Apollo Scientific

Synonyms: Acetic Acid 2-Ethylphenyl Ester

OR922066-25G OR922066-5G
APOSOR922066-25GEA 14880 CZK
APOSOR922066-25G APOSOR922066-5G
2-Ethylphenyl acetate 95%
2-Ethylphenyl acetate
Vzorec: C₁₀H₁₂O₂
MW: 164.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093429
CAS číslo: 3056-59-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR