Tisk…

4-Methoxy-2-methylphenylacetic acid 95%

Dodavatel: Apollo Scientific

OR922010-1G OR922010-250MG
APOSOR922010-1GEA 10580 CZK
APOSOR922010-1G APOSOR922010-250MG
4-Methoxy-2-methylphenylacetic acid 95%
4-Methoxy-2-methylphenylacetic acid
Vzorec: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD02664778
CAS číslo: 942-97-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR