Methyl-3-methylenecyclobutanecarboxylate
Dodavatel: Apollo Scientific
OR921971-10G
OR921971-1G
OR921971-5G
APOSOR921971-10GEA
36040
CZK
APOSOR921971-10G
APOSOR921971-1G
APOSOR921971-5G
Methyl-3-methylenecyclobutanecarboxylate
Methyl-3-methylenecyclobutanecarboxylate
Vzorec:
C₇H₁₀O₂ Storage Temperature: Ambient |
MDL Number:
MFCD01087180 CAS číslo: 15963-40-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...