Tisk…

Chloromethyl cyclohexyl ether 95%

Dodavatel: Apollo Scientific

Synonyms: (Chloromethoxy)cyclohexane

OR921970-25G
APOSOR921970-25GEA 5230 CZK
APOSOR921970-25G
Chloromethyl cyclohexyl ether 95%
Chloromethyl cyclohexyl ether
Vzorec: C₇H₁₃ClO
MW: 148,63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566884
CAS číslo: 3587-62-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR