3-Bromoisobutyric acid 95%
Dodavatel: Apollo Scientific
OR921957-25G
OR921957-5G
APOSOR921957-25GEA
11410
CZK
APOSOR921957-25G
APOSOR921957-5G
3-Bromoisobutyric acid 95%
3-Bromoisobutyric acid
Vzorec:
C₄H₇BrO₂ Storage Temperature: Ambient |
MDL Number:
MFCD01310530 CAS číslo: 56970-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...