Tisk…

4-Bromo-2-tert-butylphenol

Dodavatel: Apollo Scientific
Varování

OR921949-5G
APOSOR921949-5GEA 1950 CZK
APOSOR921949-5G
4-Bromo-2-tert-butylphenol
4-Bromo-2-tert-butylphenol
Vzorec: C₁₀H₁₃BrO
MW: 229,12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00192645
CAS číslo: 10323-39-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR