3-(Methylthio)propionyl chloride 95%
Dodavatel: Apollo Scientific
Synonyms:
3-(Methylmercapto)propionyl Chloride
OR921893-5G
APOSOR921893-5GEA
2240
CZK
APOSOR921893-5G
3-(Methylthio)propionyl chloride 95%
3-(Methylthio)propionyl chloride
Vzorec:
C₄H₇ClOS MW: 138,61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00059482 CAS číslo: 7031-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...