4-tert-Pentylcyclohexanone 95%
Dodavatel: Apollo Scientific
Synonyms:
4-(tert-Pentyl)cyclohexanone, 4-tert-Amylcyclohexanone
OR921862-100G
OR921862-25G
APOSOR921862-100GEA
4070
CZK
APOSOR921862-100G
APOSOR921862-25G
4-tert-Pentylcyclohexanone 95%
4-tert-Pentylcyclohexanone
Vzorec:
C₁₁H₂₀O MW: 168.28 Bod varu: 231 Bod tání: 96 Hustota: 0.9245 Storage Temperature: Ambient |
MDL Number:
MFCD00013798 CAS číslo: 16587-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...