3-Methylbenzyl alcohol 97%
Dodavatel: Apollo Scientific
Synonyms:
3-(Hydroxymethyl)toluole, (3-Methylphenyl)methanol, m-Tolylmethanol
OR921829-100G
APOSOR921829-100GEA
3020
CZK
APOSOR921829-100G
3-Methylbenzyl alcohol 97%
3-Methylbenzyl alcohol
Vzorec:
C₈H₁₀O MW: 122,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004646 CAS číslo: 587-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...