Tisk…
O-Butylhydroxylamine hydrochloride 98%
Dodavatel: Apollo Scientific
OR921791-5G
APOSOR921791-5GEA 2440 CZK
APOSOR921791-5G
O-Butylhydroxylamine hydrochloride 98%
O-Butylhydroxylamine hydrochloride
Vzorec: C₄H₁₁NO·HCl
MW: 125,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00487631
CAS číslo: 4490-82-8