1-Boc-4-(3-hydroxypropyl)piperazine 98%
Dodavatel: Apollo Scientific
OR921732-25G
OR921732-5G
APOSOR921732-25GEA
6220
CZK
APOSOR921732-25G
APOSOR921732-5G
1-Boc-4-(3-hydroxypropyl)piperazine 98%
1-Boc-4-(3-hydroxypropyl)piperazine
Vzorec:
C₁₂H₂₄N₂O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD06798090 CAS číslo: 132710-90-8 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...