Cyclopentanemethylamine 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR921604-1G
OR921604-25G
OR921604-5G
APOSOR921604-1GEA
3540
CZK
APOSOR921604-1G
APOSOR921604-25G
APOSOR921604-5G
Cyclopentanemethylamine 98%
Cyclopentanemethylamine
Vzorec:
C₆H₁₃N Storage Temperature: Ambient |
MDL Number:
MFCD04114081 CAS číslo: 6053-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...