Tisk…

2-Bromo-6-vinylpyridine 95%

Dodavatel: Apollo Scientific

OR921461-1G OR921461-250MG
APOSOR921461-1GEA 13160 CZK
APOSOR921461-1G APOSOR921461-250MG
2-Bromo-6-vinylpyridine 95%
2-Bromo-6-vinylpyridine
Vzorec: C₇H₆BrN
Storage Temperature: Freezer
MDL Number: MFCD18255049
CAS číslo: 931582-13-7
UN: 3073
ADR: 6.1+3+8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR