Tisk…

Isopropyl cyclohexanecarboxylate 95%

Dodavatel: Apollo Scientific

Synonyms: Cyclohexanecarboxylic Acid Isopropyl Ester

OR921446-100G OR921446-25G
APOSOR921446-100GEA 10750 CZK
APOSOR921446-100G APOSOR921446-25G
Isopropyl cyclohexanecarboxylate 95%
Isopropyl cyclohexanecarboxylate
Vzorec: C₁₀H₁₈O₂
MW: 170,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058949
CAS číslo: 6553-80-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR