Tisk…
2,3-Dimethylaniline hydrochloride 97%
Dodavatel: Apollo Scientific
Synonyms: 2,3-Xylidine Hydrochloride
OR921368-100G
APOSOR921368-100GEA 8600 CZK
APOSOR921368-100G
2,3-Dimethylaniline hydrochloride 97%
2,3-Dimethylaniline hydrochloride
Vzorec: C₈H₁₁N
MW: 157,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00054332
CAS číslo: 5417-45-8