Tisk…

tert-Butyl-3-oxocyclobutanecarboxylate 95%

Dodavatel: Apollo Scientific
Varování

OR921343-25G OR921343-10G OR921343-5G
APOSOR921343-25GEA 2440 CZK
APOSOR921343-25G APOSOR921343-10G APOSOR921343-5G
tert-Butyl-3-oxocyclobutanecarboxylate 95%
tert-Butyl-3-oxocyclobutanecarboxylate
Vzorec: C₉H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD12827541
CAS číslo: 145549-76-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR