Dimethyl(4-pyrrolidin-2-ylphenyl)aminemonoacetate 95%
Dodavatel: Apollo Scientific
Synonyms:
N,N-dimethyl-4-(pyrrolidin-2-yl)aniline
OR921273-1G
OR921273-250MG
APOSOR921273-1GEA
13230
CZK
APOSOR921273-1G
APOSOR921273-250MG
Dimethyl(4-pyrrolidin-2-ylphenyl)aminemonoacetate 95%
Dimethyl(4-pyrrolidin-2-ylphenyl)aminemonoacetate
Vzorec:
C₁₄H₂₂N₂O₂ MW: 250,34 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06660377 CAS číslo: 298690-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...