Tisk…

Benzyl formate 95%

Dodavatel: Apollo Scientific

OR921262-1KG OR921262-500G OR921262-5G OR921262-25G
APOSOR921262-1KGEA 4400 CZK
APOSOR921262-1KG APOSOR921262-500G APOSOR921262-5G APOSOR921262-25G
Benzyl formate 95%
Benzyl formate
Vzorec: C₈H₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00014128
CAS číslo: 104-57-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR