Tisk…

Methyl-2-methoxyphenylacetate 98%

Dodavatel: Apollo Scientific

Synonyms: 2-Methoxyphenylacetic Acid Methyl Ester, Methyl 2-Methoxyphenylacetate

OR921256-100G
APOSOR921256-100GEA 6090 CZK
APOSOR921256-100G
Methyl-2-methoxyphenylacetate 98%
Methyl-2-methoxyphenylacetate
Vzorec: C₁₀H₁₂O₃
MW: 180.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02093479
CAS číslo: 27798-60-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR