Tisk…

Ethyl-4-pentenoate 96%

Dodavatel: Apollo Scientific

Synonyms: Ethyl pent-4-enoate, 4-Pentenoic Acid Ethyl Ester

OR921169-100G OR921169-25G
APOSOR921169-100GEA 13790 CZK
APOSOR921169-100G APOSOR921169-25G
Ethyl-4-pentenoate 96%
Ethyl-4-pentenoate
Vzorec: C₇H₁₂O₂
MW: 128,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036562
CAS číslo: 1968-40-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR