Phenaceturic acid 98%
Dodavatel: Apollo Scientific
OR921146-25G
OR921146-100G
APOSOR921146-25GEA
3540
CZK
APOSOR921146-25G
APOSOR921146-100G
Phenaceturic acid 98%
Phenaceturic acid
Vzorec:
C₁₀H₁₁NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00021744 CAS číslo: 500-98-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...