Tisk…
S-Sodium ethanethiosulphonate 95%
Dodavatel: Apollo Scientific
Synonyms: Ethanethiosulphonic acid S-Sodium Salt
OR921031-100G OR921031-25G OR921031-5G
APOSOR921031-100GEA 12200 CZK
APOSOR921031-100G APOSOR921031-25G APOSOR921031-5G
S-Sodium ethanethiosulphonate 95%
S-Sodium ethanethiosulphonate
Vzorec: C₂H₆O₂S₂
MW: 148.17 g/mol
Storage Temperature: Ambient
CAS číslo: 31999-88-9