Tisk…

2-Hydroxy-2-methylbutyric acid 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Hydroxy-2-methylbutanoic acid

OR920941-25G
APOSOR920941-25GEA 1870 CZK
APOSOR920941-25G
2-Hydroxy-2-methylbutyric acid 95%
2-Hydroxy-2-methylbutyric acid
Vzorec: C₅H₁₀O₃
MW: 118,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004476
CAS číslo: 3739-30-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR