Tisk…

1-Methyl-2-phenylethyl acetate 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Acetoxy-1-phenylpropane, Acetic Acid 1-Methyl-2-phenylethyl Ester

OR920861-1G OR920861-5G
APOSOR920861-1GEA 5720 CZK
APOSOR920861-1G APOSOR920861-5G
1-Methyl-2-phenylethyl acetate 95%
1-Methyl-2-phenylethyl acetate
Vzorec: C₁₁H₁₄O₂
MW: 178,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01672540
CAS číslo: 2114-33-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR