Isobutyl tiglate 95%
Dodavatel: Apollo Scientific
Synonyms:
Tiglic acid Isobutyl ester
OR920788-25G
OR920788-5G
APOSOR920788-25GEA
8070
CZK
APOSOR920788-25G
APOSOR920788-5G
Isobutyl tiglate 95%
Isobutyl tiglate
Vzorec:
C₉H₁₆O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00016629 CAS číslo: 61692-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...