1-(Dimethylethylsilyl)imidazole 95%
Dodavatel: Apollo Scientific
OR920765-1G
OR920765-5G
APOSOR920765-1GEA
6620
CZK
APOSOR920765-1G
APOSOR920765-5G
1-(Dimethylethylsilyl)imidazole 95%
1-(Dimethylethylsilyl)imidazole
Vzorec:
C₇H₁₄N₂Si Storage Temperature: Ambient |
MDL Number:
MFCD00059723 CAS číslo: 62365-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...