(±)-3,3-Dimethyl-2-butylamine 95%
Dodavatel: Apollo Scientific
Synonyms:
1,2,2-Trimethylpropylamine, 3,3-Dimethylbutan-2-amine
OR920743-1G
OR920743-5G
APOSOR920743-1GEA
2440
CZK
APOSOR920743-1G
APOSOR920743-5G
(±)-3,3-Dimethyl-2-butylamine 95%
(±)-3,3-Dimethyl-2-butylamine
Vzorec:
C₆H₁₅N MW: 101,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00008078 CAS číslo: 3850-30-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...