Tisk…

4-Propylaniline 95%

Dodavatel: Apollo Scientific

OR920683-25G OR920683-5G
APOSOR920683-25GEA 8100 CZK
APOSOR920683-25G APOSOR920683-5G
4-Propylaniline 95%
4-Propylaniline
Vzorec: C₉H₁₃N
Storage Temperature: Ambient
MDL Number: MFCD00007924
CAS číslo: 2696-84-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR