Tisk…

2-(Chloromethyl)-3,5-dioxahex-1-ene 95%

Dodavatel: Apollo Scientific

Synonyms: 3-Chloro-2-(methoxymethoxy)-1-propene

OR920588-1G OR920588-5G
APOSOR920588-1GEA 6880 CZK
APOSOR920588-1G APOSOR920588-5G
2-(Chloromethyl)-3,5-dioxahex-1-ene 95%
2-(Chloromethyl)-3,5-dioxahex-1-ene
Vzorec: C₅H₉ClO₂
MW: 136,58 g/mol
Bod varu: 63 °C
Hustota: 1,1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00130115
CAS číslo: 105104-40-3
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR