Tisk…

4-Ethylcyclohexanol 95%

Dodavatel: Apollo Scientific

OR920372-25G
APOSOR920372-25GEA 1820 CZK
APOSOR920372-25G
4-Ethylcyclohexanol 95%
4-Ethylcyclohexanol
Vzorec: C₈H₁₆O
MW: 128,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001450
CAS číslo: 4534-74-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR