2,4-Dichloro-6-methoxy-1,3,5-triazine 95%
Dodavatel: Apollo Scientific
OR920230-10G
OR920230-25G
APOSOR920230-10GEA
1330
CZK
APOSOR920230-10G
APOSOR920230-25G
2,4-Dichloro-6-methoxy-1,3,5-triazine 95%
2,4-Dichloro-6-methoxy-1,3,5-triazine
Vzorec:
C₄H₃Cl₂N₃O MW: 179,9921 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00128159 CAS číslo: 3638-04-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...