Tisk…
sec-Butyl crotonate 95%
Dodavatel: Apollo Scientific
Synonyms: Crotonic Acid sec-Butyl Ester
OR920185-100G OR920185-25G OR920185-5G
APOSOR920185-100GEA 16340 CZK
APOSOR920185-100G APOSOR920185-25G APOSOR920185-5G
sec-Butyl crotonate 95%
sec-Butyl crotonate
Vzorec: C₈H₁₄O₂
MW: 142.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059054
CAS číslo: 10371-45-6