3-Buten-2-ylacetate 95%
Dodavatel: Apollo Scientific
Synonyms:
3-Acetoxy-1-butene, 1-Methylallyl Acetate, Acetic Acid 3-Buten-2-yl Ester, But-3-en-2-yl acetate
OR920138-5G
APOSOR920138-5GEA
5690
CZK
APOSOR920138-5G
3-Buten-2-ylacetate 95%
3-Buten-2-ylacetate
Vzorec:
C₆H₁₀O₂ MW: 114,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00026202 CAS číslo: 6737-11-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...